2-(Chloromethyl)-3,4-dimethoxypyridine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Chloromethyl-3,4-dimethoxypyridinium chloride, 2-(Chloromethyl)-3,4-dimethoxypyridine HCl

C2073-25G C2073-5G
TCC2073-25GEA 195 USD
TCC2073-25G TCC2073-5G
2-(Chloromethyl)-3,4-dimethoxypyridine hydrochloride ≥98.0% (by titrimetric analysis)
2-(Chloromethyl)-3,4-dimethoxypyridine hydrochloride

CAS Number: 72830-09-2
MDL Number: MFCD02181083
Molecular Formula: C8H10ClNO2
Molecular Weight: 224.08
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 150

Formula: C₈H₁₀ClNO₂
MW: 224.08 g/mol
Melting Pt: 150 °C
MDL Number: MFCD02181083
CAS Number: 72830-09-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=90f28c1a329802eb17cedfe8b5ae812717abaf6e&vid=90f28c1a329802eb17cedfe8b5ae812717abaf6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR