3-Chlorothioanisole ≥96.0% (by GC)

Supplier: TCI America

Synonyms: (3-Chlorophenyl)(methyl)sulphane, 3-Chlorophenyl methyl sulphide, 1-Chloro-3-(methylthio)benzene

C2070-25G C2070-5G
TCC2070-25GEA 60.7 USD
TCC2070-25G TCC2070-5G
3-Chlorothioanisole ≥96.0% (by GC)
3-Chlorothioanisole

CAS Number: 4867-37-2
MDL Number: MFCD00040127
Molecular Formula: C7H7ClS
Molecular Weight: 158.64
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 244
Flash Point (°C): 106
Specific Gravity (20/20): 1.22


Other Product Names / Synonyms: 3-Chlorophenyl Methyl Sulfide, 1-Chloro-3-(methylthio)benzene

Formula: C₇H₇ClS
MW: 158.65 g/mol
Boiling Pt: 244 °C
Density: 1.22 (20 °C)
Flash Pt: 106 °C
MDL Number: MFCD00040127
CAS Number: 4867-37-2
UN: 3334
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=01007a503cd3132bd2db904097f596eec529aad3&vid=01007a503cd3132bd2db904097f596eec529aad3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR