2-Chlorothioanisole ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1-Chloro-2-(methylthio)benzene, 2-Chlorophenyl methyl sulphide
C2069-25G
C2069-5G
TCC2069-25GEA
63
USD
TCC2069-25G
TCC2069-5G
2-Chlorothioanisole ≥98.0% (by GC)
2-Chlorothioanisole
CAS Number: 17733-22-1
MDL Number: MFCD00040124
Molecular Formula: C7H7ClS
Molecular Weight: 158.64
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 227
Specific Gravity (20/20): 1.24
Other Product Names / Synonyms: 2-Chlorophenyl Methyl Sulfide, 1-Chloro-2-(methylthio)benzene
Formula:
C₇H₇ClS MW: 158.65 g/mol Boiling Pt: 227 °C Density: 1.24 (20 °C) |
MDL Number:
MFCD00040124 CAS Number: 17733-22-1 UN: 3334 ADR: 9, |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...