2-Chlorobenzyl mercaptan ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Chlorobenzenemethanethiol, o-Chlorotoluene-α-thiol
C2068-25G
C2068-5G
TCC2068-25GEA
68
USD
TCC2068-25G
TCC2068-5G
2-Chlorobenzyl mercaptan ≥98.0% (by GC)
2-Chlorobenzyl mercaptan
CAS Number: 39718-00-8
MDL Number: MFCD00004868
Molecular Formula: C7H7ClS
Molecular Weight: 158.64
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 121
Flash Point (°C): 69
Specific Gravity (20/20): 1.23
Other Product Names / Synonyms: 2-Chlorobenzenemethanethiol, (2-Chlorophenyl)methanethiol
Formula:
C₇H₇ClS MW: 158.65 g/mol Boiling Pt: 121 °C Density: 1.23 (20 °C) Flash Pt: 69 °C |
MDL Number:
MFCD00004868 CAS Number: 39718-00-8 UN: 2810 ADR: 6.1,III |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...