4-Chlorothiobenzamide ≥97.0% (by HPLC, total nitrogen)

Supplier: TCI America

Synonyms: 4-Chlorobenzene-1-carbothioamide

C2053-25G C2053-5G
TCC2053-25GEA 59 USD
TCC2053-25G TCC2053-5G
4-Chlorothiobenzamide ≥97.0% (by HPLC, total nitrogen)
4-Chlorothiobenzamide

CAS Number: 2521-24-6
Molecular Formula: C7H6ClNS
Molecular Weight: 171.64
Purity/Analysis Method: >97.0% (HPLC,N)
Form: Crystal
Melting point (°C): 129

Formula: C₇H₆ClNS
MW: 171.65 g/mol
Melting Pt: 129 °C
MDL Number: MFCD00040956
CAS Number: 2521-24-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa518c6d639a58c2cd1c9f591b5b1e00533f0afd&vid=fa518c6d639a58c2cd1c9f591b5b1e00533f0afd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (HPLC,N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR