4-Cyano-2-fluorobenzoic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C2051-1G C2051-5G
TCC2051-1GEA 28.2 USD
TCC2051-1G TCC2051-5G
4-Cyano-2-fluorobenzoic acid ≥97.0% (by GC, titration analysis)
4-Cyano-2-fluorobenzoic acid

CAS Number: 164149-28-4
MDL Number: MFCD03094454
Molecular Formula: C8H4FNO2
Molecular Weight: 165.12
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal
Melting point (°C): 216

Formula: C₈H₄FNO₂
MW: 165.12 g/mol
Melting Pt: 216 °C
MDL Number: MFCD03094454
CAS Number: 164149-28-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9fa2fe9694caafc48f3e5510a565b19ccfda8d00&vid=9fa2fe9694caafc48f3e5510a565b19ccfda8d00&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR