Caffeine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: 3,7-Dihydro-1,3,7-trimethyl-1H-purine-2,6-dione, 7-Methyltheobromine, 1,3,7-Trimethylxanthine, Methyltheobromine, 1,3,7-Trimethyl-2,6-dioxopurine, 2,6-Dihydroxy-1,3,7-trimethylpurine

C2042-25G C2042-500G
TCC2042-25GEA 34.23 USD
TCC2042-25G TCC2042-500G
Caffeine ≥98.0% (by HPLC, titration analysis)
Caffeine

CAS Number: 58-08-2
Molecular Formula: C8H10N4O2
Molecular Weight: 194.19
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 238


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Crude Medicine Ingredients for Research and Experimental Use, Alkaloids (Phytochemicals), Nucleobases and Their Analogs, Purine Alkaloids
Other Product Names / Synonyms: 1,3,7-Trimethylxanthine

Formula: C₈H₁₀N₄O₂
MW: 194.19 g/mol
Melting Pt: 238 °C
MDL Number: MFCD00005758
CAS Number: 58-08-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7ed7537432e5e118c5cd0792258523e7a16a66&vid=28e0e3c3e9f28e75b14f65379283ded6efc2d5fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR