ω-Chloro-2,4-difluoroacetophenone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-Chloro-1-(2,4-difluorophenyl)ethanone, α-Chloro-2,4-difluoroacetophenone, 2-Chloro-2',4'-difluoroacetophenone

C2030-25G C2030-5G
TCC2030-25GEA 110 USD
TCC2030-25G TCC2030-5G
ω-Chloro-2,4-difluoroacetophenone ≥98.0% (by GC)
ω-Chloro-2,4-difluoroacetophenone

CAS Number: 51336-94-8
Molecular Formula: C8H5ClF2O
Molecular Weight: 190.57
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 50
Flash Point (°C): 110


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2,4-Difluorophenacyl Chloride

Formula: C₈H₅ClF₂O
MW: 190.58 g/mol
Melting Pt: 50 °C
Flash Pt: 110 °C
MDL Number: MFCD00013252
CAS Number: 51336-94-8
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05d0cdccdb3cf0cb1763d30fa57ca5d5bcb029b0&vid=05d0cdccdb3cf0cb1763d30fa57ca5d5bcb029b0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR