2-Chloro-p-cresol ≥97.0%

Supplier: TCI America

Synonyms: 1-Chloro-2-hydroxy-5-methylbenzene, 2-Chloro-4-methylphenol

C2005-25G C2005-5G
TCC2005-25GEA 111 USD
TCC2005-25G TCC2005-5G
2-Chloro-p-cresol ≥97.0%
2-Chloro-p-cresol

CAS Number: 6640-27-3
Molecular Formula: C7H7ClO
Molecular Weight: 142.58
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 196
Flash Point (°C): 86
Specific Gravity (20/20): 1.21


Other Product Names / Synonyms: 2-Chloro-4-methylphenol

Formula: C₇H₇ClO
Boiling Pt: 196 °C
Density: 1.21 (20 °C)
Flash Pt: 86 °C
MDL Number: MFCD00070639
CAS Number: 6640-27-3
UN: 2669
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8ec814d8bdff70aebd023e371563ed60a87aaf1&vid=a8ec814d8bdff70aebd023e371563ed60a87aaf1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR