2-Chloro-6-(trifluoromethyl)pyridine ≥98.0%

Supplier: TCI America

C1986-1G C1986-5G
TCC1986-1GEA 41 USD
TCC1986-1G TCC1986-5G
2-Chloro-6-(trifluoromethyl)pyridine ≥98.0%
2-Chloro-6-(trifluoromethyl)pyridine

CAS Number: 39890-95-4
Molecular Formula: C6H3ClF3N
Molecular Weight: 181.54
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 78
Flash Point (°C): 79
Freezing point (°C): 33

Formula: C₆H₃ClF₃N
MW: 181.54
Boiling Pt: 78 °C
Flash Pt: 79 °C
MDL Number: MFCD00042226
CAS Number: 39890-95-4
UN: 2811
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bd30824923bc7f941eda9a8cca1210a97852a428&vid=bd30824923bc7f941eda9a8cca1210a97852a428&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR