2-Cyanoethyl acrylate ≥95.0% (by GC) stabilized
Supplier: TCI America
(stabilized with MEHQ)
CAS Number: 106-71-8
Molecular Formula: C6H7NO2
Molecular Weight: 125.13
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 108
Flash Point (°C): 124
Specific Gravity (20/20): 1.07
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Acrylic Acid 2-Cyanoethyl Ester, 3-Acryloyloxypropionitrile
Formula:
C₆H₇NO₂ Boiling Pt: 108 °C Density: 1.07 (20 °C) Flash Pt: 124 °C |
MDL Number:
MFCD00013823 CAS Number: 106-71-8 |
|
Specification Test Results
Purity/Analysis Method | >95.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...