Cyclobutanone ≥97.0% (by GC) stabilized

Supplier: TCI America

C1913-1G C1913-5G
TCC1913-1GEA 56 USD
TCC1913-1G TCC1913-5G
Cyclobutanone ≥97.0% (by GC) stabilized
Cyclobutanone

(stabilized with Na2CO3)
CAS Number: 1191-95-3
MDL Number: MFCD00001332
Molecular Formula: C4H6O
Molecular Weight: 70.09
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 99
Melting point (°C): -53
Flash Point (°C): 10
Specific Gravity (20/20): 0.93
Storage Temperature: 0-10°C

Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 99 °C
Melting Pt: –53 °C
Density: 0.93 (20 °C)
Flash Pt: 10 °C
MDL Number: MFCD00001332
CAS Number: 1191-95-3
UN: 1224
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c87df8957053738d3a53d7fd954dc66947bbeeee&vid=fbf2c0aa5ab86f74b1393b8e26f6d8df0ac3b81f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR