4-Chloro-3-nitrobenzenesulfonyl chloride ≥96.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4-Chloro-3-nitrobenzenesulphonyl Chloride, 4-Chloro-3-nitrobenzene-1-sulphonyl chloride

C1887-25G C1887-5G
TCC1887-25GEA 312.8 USD
TCC1887-25G TCC1887-5G
4-Chloro-3-nitrobenzenesulfonyl chloride ≥96.0% (by GC, titration analysis)
4-Chloro-3-nitrobenzenesulphonyl chloride

CAS Number: 97-08-5
Molecular Formula: C6H3Cl2NO4S
Molecular Weight: 256.05
Purity/Analysis Method: >96.0% (GC,T)
Form: Crystal
Melting point (°C): 60

Formula: C₆H₃Cl₂NO₄S
MW: 256.07 g/mol
Melting Pt: 60 °C
MDL Number: MFCD00007440
CAS Number: 97-08-5
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e269d3f77425653e78604a83a700a44e65efdf4&vid=3e269d3f77425653e78604a83a700a44e65efdf4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >96.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR