4-Cyanobenzenesulphonyl chloride ≥98.0%
Supplier: TCI America
Synonyms:
4-Cyanobenzene-1-sulphonyl chloride
C1813-1G
C1813-5G
TCC1813-1GEA
32
USD
TCC1813-1G
TCC1813-5G
4-Cyanobenzenesulphonyl chloride ≥98.0%
4-Cyanobenzenesulphonyl chloride
CAS Number: 49584-26-1
Molecular Formula: C7H4ClNO2S
Molecular Weight: 201.62
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 112
Formula:
C₇H₄ClNO₂S Melting Pt: 112 °C |
MDL Number:
MFCD00052337 CAS Number: 49584-26-1 UN: 3261 ADR: 8,II |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...