3-Cyanobenzenesulphonyl chloride ≥98.0%

Supplier: TCI America

Synonyms: 3-Cyanobenzene-1-sulphonyl chloride, 3-Cyanobenzene-1-sulphonylchloride

C1812-1G C1812-5G
TCC1812-1GEA 154 USD
TCC1812-1G TCC1812-5G
3-Cyanobenzenesulphonyl chloride ≥98.0%
3-Cyanobenzenesulphonyl chloride

CAS Number: 56542-67-7
Molecular Formula: C7H4ClNO2S
Molecular Weight: 201.62
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 111
Melting point (°C): 51
Flash Point (°C): 110

Formula: C₇H₄ClNO₂S
MW: 201.63 g/mol
Boiling Pt: 111 °C
Melting Pt: 51 °C
Flash Pt: 110 °C
MDL Number: MFCD00203480
CAS Number: 56542-67-7
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0af452e480b4881b2d12e92ea1d7bcd3e2aac538&vid=0af452e480b4881b2d12e92ea1d7bcd3e2aac538&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR