2-Cyanobenzenesulphonyl chloride ≥97.0%

Supplier: TCI America

Synonyms: 2-Cyanobenzene-1-sulphonyl chloride

C1811-1G C1811-5G
TCC1811-1GEA 64 USD
TCC1811-1G TCC1811-5G
2-Cyanobenzenesulphonyl chloride ≥97.0%
2-Cyanobenzenesulphonyl chloride

CAS Number: 69360-26-5
Molecular Formula: C7H4ClNO2S
Molecular Weight: 201.62
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 68

Formula: C₇H₄ClNO₂S
MW: 201.63
Melting Pt: 67.5
MDL Number: MFCD00052341
CAS Number: 69360-26-5
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dcfd428d463944611a083bf6eed37ccf3f2ae709&vid=dcfd428d463944611a083bf6eed37ccf3f2ae709&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR