Ammonium cerium(IV) nitrate ≥98.0%

Supplier: TCI America

C1806-500G C1806-50G
TCC1806-500GEA 212.57 USD
TCC1806-500G TCC1806-50G
Ammonium cerium(IV) nitrate ≥98.0%
Ammonium cerium(IV) nitrate

CAS Number: 16774-21-3
MDL Number: MFCD00151121
Molecular Formula: H8CeN8O18
Molecular Weight: 548.22
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Oxidation, Nitration, Glycosylation
Other Product Names / Synonyms: Ceric Ammonium Nitrate, CAN, Cerium(IV) Diammonium Nitrate, Diammonium Cerium(IV) Nitrate

Formula: (NH₄)₂Ce(NO₃)₆
MDL Number: MFCD00151121
CAS Number: 16774-21-3
UN: 1477
ADR: 5.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c7323099461f824db4b19d2729b0dee9ea6fb1f4&vid=c7323099461f824db4b19d2729b0dee9ea6fb1f4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR