2-Chloro-3-methylthiophene ≥98.0% (by GC)

Supplier: TCI America

C1801-25G C1801-5G
TCC1801-25GEA 98.3 USD
TCC1801-25G TCC1801-5G
2-Chloro-3-methylthiophene ≥98.0% (by GC)
2-Chloro-3-methylthiophene

CAS Number: 14345-97-2
MDL Number: MFCD00130083
Molecular Formula: C5H5ClS
Molecular Weight: 132.61
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 54
Specific Gravity (20/20): 1.22

Formula: C₅H₅ClS
MW: 132.61 g/mol
Boiling Pt: 54 °C
Density: 1.22 (20 °C)
MDL Number: MFCD00130083
CAS Number: 14345-97-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ca57f56b35c13cb84006c0feaadda7e969e7e97a&vid=ca57f56b35c13cb84006c0feaadda7e969e7e97a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR