4-Chloro-2-iodoaniline ≥98.0% (by GC)

Supplier: TCI America

C1800-25G C1800-5G
TCC1800-25GEA 303.4 USD
TCC1800-25G TCC1800-5G
4-Chloro-2-iodoaniline ≥98.0% (by GC)
4-Chloro-2-iodoaniline

CAS Number: 63069-48-7
MDL Number: MFCD01863737
Molecular Formula: C6H5ClIN
Molecular Weight: 253.47
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 41
Flash Point (°C): 109

Formula: C₆H₅ClIN
MW: 253.47 g/mol
Melting Pt: 41 °C
Flash Pt: 109 °C
MDL Number: MFCD01863737
CAS Number: 63069-48-7
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cbaa7222e9aec57a3c8d1c4954a4110a4f2cbae5&vid=510f92717df7fcb5f04f98d8714d9c7b9d5c53d3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR