Ceric ammonium molybdate in solution (contains H2SO4)
Supplier: TCI America
Synonyms:
CAM Solution
C1794-500ML
C1794-100ML
TCC1794-500MLEA
379.47
USD
TCC1794-500ML
TCC1794-100ML
Ceric ammonium molybdate in solution (contains H2SO4)
Ceric ammonium molybdate
[for TLC Stain]
MDL Number: MFCD06797081
Form: Clear Liquid
Color: Yellow
Other Product Names / Synonyms: CAM Solution
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...