4-(Chloromethyl)styrene ≥90.0% (by GC) stabilized
Supplier: TCI America
Synonyms:
1-(Chloromethyl)-4-vinylbenzene, 4-Vinylbenzyl Chloride
C1792-250G
C1792-25G
TCC1792-250GEA
282.5
USD
TCC1792-250G
TCC1792-25G
4-(Chloromethyl)styrene ≥90.0% (by GC) stabilized
4-(Chloromethyl)styrene
(stabilized with TBC + ONP + 2-Nitro-p-cresol)
CAS Number: 1592-20-7
MDL Number: MFCD00051362
Molecular Formula: C9H9Cl
Molecular Weight: 152.62
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 229
Flash Point (°C): 103
Specific Gravity (20/20): 1.08
Storage Temperature: <0°C
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4-Vinylbenzyl Chloride
Formula:
C₉H₉Cl MW: 152.62 g/mol Boiling Pt: 229 °C Density: 1.08 (20 °C) Flash Pt: 103 °C |
MDL Number:
MFCD00051362 CAS Number: 1592-20-7 |
Specification Test Results
Purity/Analysis Method | >90.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...