2-Chloro-4-(trifluoromethyl)benzenesulfonyl chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Chloro-4-(trifluoromethyl)benzene-1-sulphonyl chloride

C1790-1G C1790-5G
TCC1790-1GEA 79.5 USD
TCC1790-1G TCC1790-5G
2-Chloro-4-(trifluoromethyl)benzenesulfonyl chloride ≥98.0% (by titrimetric analysis)
2-Chloro-4-(trifluoromethyl)benzenesulphonyl chloride

CAS Number: 175205-54-6
MDL Number: MFCD00052912
Molecular Formula: C7H3Cl2F3O2S
Molecular Weight: 279.05
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.64

Formula: C₇H₃Cl₂F₃O₂S
MW: 279.07 g/mol
Density: 1.64 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00052912
CAS Number: 175205-54-6
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=40eb8dd1f01e20ceeeb1fdef3e810b33dd45e171&vid=40eb8dd1f01e20ceeeb1fdef3e810b33dd45e171&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR