1-Chloroethylisopropyl carbonate ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
Carbonic Acid 1-Chloroethyl Isopropyl Ester, 1-Chloroethyl isopropyl carbonate
C1787-250G
C1787-25G
TCC1787-250GEA
188.3
USD
TCC1787-250G
TCC1787-25G
1-Chloroethylisopropyl carbonate ≥98.0% (by GC)
1-Chloroethylisopropyl carbonate
CAS Number: 98298-66-9
MDL Number: MFCD04038100
Molecular Formula: C6H11ClO3
Molecular Weight: 166.60
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 70
Specific Gravity (20/20): 1.09
Other Product Names / Synonyms: Carbonic Acid 1-Chloroethyl Isopropyl Ester
Formula:
C₆H₁₁ClO₃ MW: 166.60 g/mol Boiling Pt: 70 °C Density: 1.09 (20 °C) |
MDL Number:
MFCD04038100 CAS Number: 98298-66-9 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...