1-Chloroethylcyclohexyl carbonate ≥98.0% (by GC)

Supplier: TCI America

C1786-250G C1786-25G
TCC1786-250GEA 122.4 USD
TCC1786-250G TCC1786-25G
1-Chloroethylcyclohexyl carbonate ≥98.0% (by GC)
1-Chloroethylcyclohexyl carbonate

CAS Number: 99464-83-2
MDL Number: MFCD04038149
Molecular Formula: C9H15ClO3
Molecular Weight: 206.67
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 107
Flash Point (°C): 84
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: Carbonic Acid 1-Chloroethyl Cyclohexyl Ester

Formula: C₉H₁₅ClO₃
Boiling Pt: 107 °C
Density: 1.13 (20 °C)
Flash Pt: 84 °C
MDL Number: MFCD04038149
CAS Number: 99464-83-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9932ddaf419c698f946554daa39276b4b1c56837&vid=9932ddaf419c698f946554daa39276b4b1c56837&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR