4-Chloro-m-anisidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 5-Amino-2-chloroanisole, 4-Chloro-3-methoxyaniline, 4-Chloro-3-anisidine

C1774-1G C1774-5G
TCC1774-1GEA 35 USD
TCC1774-1G TCC1774-5G
4-Chloro-m-anisidine ≥98.0% (by GC, titration analysis)
4-Chloro-m-anisidine

CAS Number: 13726-14-2
MDL Number: MFCD00672967
Molecular Formula: C7H8ClNO
Molecular Weight: 157.60
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 79


Other Product Names / Synonyms: 4-Chloro-m-anisidine, 5-Amino-2-chloroanisole, 2-Chloro-5-aminoanisole

Formula: C₇H₈ClNO
MW: 157.6 g/mol
Melting Pt: 79 °C
MDL Number: MFCD00672967
CAS Number: 13726-14-2
UN: 2233
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0279376c19264565cbd5ba81f7f10ac9676e1ba0&vid=0279376c19264565cbd5ba81f7f10ac9676e1ba0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR