3-Chloro-4-fluorobenzenesulphonyl chloride ≥98.0%

Supplier: TCI America

Synonyms: 3-Chloro-4-fluorobenzene-1-sulphonyl chloride

C1772-25G C1772-5G
TCC1772-25GEA 169.5 USD
TCC1772-25G TCC1772-5G
3-Chloro-4-fluorobenzenesulphonyl chloride ≥98.0%
3-Chloro-4-fluorobenzenesulphonyl chloride

CAS Number: 91170-93-3
MDL Number: MFCD00041444
Molecular Formula: C6H3Cl2FO2S
Molecular Weight: 229.05
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.59

Formula: C₆H₃Cl₂FO₂S
Density: 1.59 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00041444
CAS Number: 91170-93-3
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=acc4160e3b31e641afa0fa1731f7bd6582446500&vid=acc4160e3b31e641afa0fa1731f7bd6582446500&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR