Methyl-4-(chloromethyl)benzoate ≥98.0% (by GC)

Supplier: TCI America

C1763-25G C1763-5G
TCC1763-25GEA 221.8 USD
TCC1763-25G TCC1763-5G
Methyl-4-(chloromethyl)benzoate ≥98.0% (by GC)
Methyl 4-(chloromethyl)benzoate

CAS Number: 34040-64-7
MDL Number: MFCD00511224
Molecular Formula: C9H9ClO2
Molecular Weight: 184.62
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 165
Melting point (°C): 39
Storage Temperature: 0-10°C


Other Product Names / Synonyms: alpha-Chloro-p-toluic Acid Methyl Ester, 4-(Chloromethyl)benzoic Acid Methyl Ester, Methyl alpha-Chloro-p-toluate

Formula: C₉H₉ClO₂
Boiling Pt: 165 °C
Melting Pt: 39 °C
MDL Number: MFCD00511224
CAS Number: 34040-64-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2acd55ce6f84e516891797426242c267b1c92f12&vid=2acd55ce6f84e516891797426242c267b1c92f12&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR