5-Chloro-o-cresol ≥97.0%

Supplier: TCI America

Synonyms: 1-Chloro-3-hydroxy-4-methylbenzene, 5-Chloro-2-methylphenol

C1723-1G C1723-5G
TCC1723-1GEA 30.08 USD
TCC1723-1G TCC1723-5G
5-Chloro-o-cresol ≥97.0%
5-Chloro-o-cresol

CAS Number: 5306-98-9
MDL Number: MFCD00060672
Molecular Formula: C7H7ClO
Molecular Weight: 142.58
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 74


Other Product Names / Synonyms: 4-Chloro-2-hydroxytoluene, 5-Chloro-2-methylphenol

Formula: C₇H₇ClO
Melting Pt: 74 °C
MDL Number: MFCD00060672
CAS Number: 5306-98-9
UN: 3437
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a439ae8d4333e75787973c643f79f3acae02f147&vid=a439ae8d4333e75787973c643f79f3acae02f147&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR