4,5,6,7-Tetrahydroisobenzofuran-1,3-dione ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Cyclohexene-1,2-dicarboxylic anhydride, 3,4,5,6-Tetrahydrophthalic anhydride

C1701-25G C1701-5G
TCC1701-25GEA 43 USD
TCC1701-25G TCC1701-5G
4,5,6,7-Tetrahydroisobenzofuran-1,3-dione ≥98.0% (by GC)
4,5,6,7-Tetrahydroisobenzofuran-1,3-dione

CAS Number: 2426-02-0
MDL Number: MFCD00005917
Molecular Formula: C8H8O3
Molecular Weight: 152.15
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 72


Other Product Names / Synonyms: 3,4,5,6-Tetrahydrophthalic Anhydride

Formula: C₈H₈O₃
MW: 152.15
Melting Pt: 72 °C
MDL Number: MFCD00005917
CAS Number: 2426-02-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e182b649a301d1cf328117c30a2add2f89807742&vid=e182b649a301d1cf328117c30a2add2f89807742&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR