2-Chloro-3-methoxypropionic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
C1688-25G
C1688-5G
TCC1688-25GEA
690
USD
TCC1688-25G
TCC1688-5G
2-Chloro-3-methoxypropionic acid ≥98.0% (by GC, titration analysis)
2-Chloro-3-methoxypropionic acid
CAS Number: 2544-05-0
MDL Number: MFCD03093624
Molecular Formula: C4H7ClO3
Molecular Weight: 138.55
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 155
Specific Gravity (20/20): 1.30
Formula:
C₄H₇ClO₃ Boiling Pt: 155 °C Density: 1.30 (20 °C) |
MDL Number:
MFCD03093624 CAS Number: 2544-05-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...