1-Chloro-2-butene (mixture of cis and trans isomers) ≥70.0% (by GC)
Supplier: TCI America
Synonyms:
Crotyl chloride, 1-Chloro-2-butene, 2-Buten-1-yl chloride
C1686-25ML
C1686-500ML
TCC1686-25MLEA
61.33
USD
TCC1686-25ML
TCC1686-500ML
1-Chloro-2-butene (mixture of cis and trans isomers) ≥70.0% (by GC)
1-Chloro-2-butene (mixture of cis and trans isomers)
CAS Number: 591-97-9
MDL Number: MFCD00000987
Molecular Formula: C4H7Cl
Molecular Weight: 90.55
Purity/Analysis Method: >70.0% (GC)
Form: Clear Liquid
Flash Point (°C): -10
Specific Gravity (20/20): 0.93
Other Product Names / Synonyms: 2-Buten-1-yl Chloride, 1-Chloro-2-butene
Formula:
C₄H₇Cl MW: 90.55 g/mol Density: 0.93 (20 °C) Flash Pt: -10 °C |
MDL Number:
MFCD00000987 CAS Number: 591-97-9 |
Specification Test Results
Purity/Analysis Method | >70.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...