α-Bromo-m-toluonitrile ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3-(Bromomethyl)benzonitrile, m-(Bromomethyl)benzonitrile

C1665-25G C1665-5G
TCC1665-25GEA 140 USD
TCC1665-25G TCC1665-5G
α-Bromo-m-toluonitrile ≥98.0% (by GC)
α-Bromo-m-toluonitrile

CAS Number: 28188-41-2
MDL Number: MFCD00001809
Molecular Formula: C8H6BrN
Molecular Weight: 196.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 130
Melting point (°C): 95


Other Product Names / Synonyms: alpha-Bromo-m-tolunitrile, 3-Cyanobenzyl Bromide

Formula: C₈H₆BrN
MW: 196.05 g/mol
Boiling Pt: 130 °C
Melting Pt: 95 °C
MDL Number: MFCD00001809
CAS Number: 28188-41-2
UN: 2923
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9db5ca27c615c1daaa22ae8d25a066d6fc803ed5&vid=9db5ca27c615c1daaa22ae8d25a066d6fc803ed5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR