3-Hydroxypyridine-2-carbonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Cyano-3-hydroxypyridine, 3-Hydroxypicolinonitrile

C1664-25G C1664-5G
TCC1664-25GEA 970.8 USD
TCC1664-25G TCC1664-5G
3-Hydroxypyridine-2-carbonitrile ≥98.0% (by GC, titration analysis)
3-Hydroxypyridine-2-carbonitrile

CAS Number: 932-35-4
MDL Number: MFCD01646135
Molecular Formula: C6H4N2O
Molecular Weight: 120.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal


Other Product Names / Synonyms: 3-Hydroxypicolinonitrile

Formula: C₆H₄N₂O
MW: 120.1088 g/mol
MDL Number: MFCD01646135
CAS Number: 932-35-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=842c117014453420dd38353edd5c5441c01a7fcd&vid=842c117014453420dd38353edd5c5441c01a7fcd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR