2-Furonitrile ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
2-Cyanofuran
C1648-25G
C1648-5G
TCC1648-25GEA
310
USD
TCC1648-25G
TCC1648-5G
2-Furonitrile ≥98.0% (by GC)
2-Furonitrile
CAS Number: 617-90-3
MDL Number: MFCD00003223
Molecular Formula: C5H3NO
Molecular Weight: 93.09
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 148
Flash Point (°C): 35
Specific Gravity (20/20): 1.08
Product Categories: Synthetic Organic Chemistry, Building Blocks ([2+2+2] Cycloaddition Reaction)
Other Product Names / Synonyms: 2-Furancarbonitrile, 2-Cyanofuran
Formula:
C₅H₃NO MW: 93.08 g/mol Boiling Pt: 148 °C Density: 1.08 (20 °C) Flash Pt: 35 °C |
MDL Number:
MFCD00003223 CAS Number: 617-90-3 UN: 1992 ADR: 3,III |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...