2-Chloro-5-(chloromethyl)pyridine ≥98.0% (by GC)

Supplier: TCI America

C1628-25G C1628-5G
TCC1628-25GEA 896 USD
TCC1628-25G TCC1628-5G
2-Chloro-5-(chloromethyl)pyridine ≥98.0% (by GC)
2-Chloro-5-(chloromethyl)pyridine

CAS Number: 70258-18-3
MDL Number: MFCD00125366
Molecular Formula: C6H5Cl2N
Molecular Weight: 162.01
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 39

Formula: C₆H₅Cl₂N
MW: 162.02 g/mol
Melting Pt: 39 °C
MDL Number: MFCD00125366
CAS Number: 70258-18-3
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=81daab53c716028952a3a770f421014236368305&vid=81daab53c716028952a3a770f421014236368305&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR