4-Chloro-N-methylaniline ≥97.0% (by GC, titration analysis)

Supplier: TCI America

C1620-25G C1620-5G
TCC1620-25GEA 362 USD
TCC1620-25G TCC1620-5G
4-Chloro-N-methylaniline ≥97.0% (by GC, titration analysis)
4-Chloro-N-methylaniline

CAS Number: 932-96-7
MDL Number: MFCD00000614
Molecular Formula: C7H8ClN
Molecular Weight: 141.60
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 142
Flash Point (°C): 51
Specific Gravity (20/20): 1.16

Formula: C₇H₈ClN
MW: 141.6
Boiling Pt: 142 °C
Density: 1.169
Flash Pt: 51 °C
MDL Number: MFCD00000614
CAS Number: 932-96-7
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ead621f05377c3b6fb7e630348d805117cd08113&vid=ead621f05377c3b6fb7e630348d805117cd08113&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR