2-Chloronicotinoyl chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America

C1604-25G C1604-5G
TCC1604-25GEA 210.3 USD
TCC1604-25G TCC1604-5G
2-Chloronicotinoyl chloride ≥98.0% (by titrimetric analysis)
2-Chloronicotinoyl chloride

CAS Number: 49609-84-9
MDL Number: MFCD00051677
Molecular Formula: C6H3Cl2NO
Molecular Weight: 176.00
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 39
Flash Point (°C): 113
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Chloropyridine-3-carbonyl Chloride

Formula: C₆H₃Cl₂NO
Boiling Pt: 120 °C
Melting Pt: 39 °C
Flash Pt: 113 °C
MDL Number: MFCD00051677
CAS Number: 49609-84-9
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e490e8a66219e5715fb057cbe0f6f34c9b979c50&vid=e490e8a66219e5715fb057cbe0f6f34c9b979c50&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR