5-Chloro-2-mercaptobenzimidazole ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Chloro-2-benzimidazolethiol

C1590-25G C1590-5G
TCC1590-25GEA 372 USD
TCC1590-25G TCC1590-5G
5-Chloro-2-mercaptobenzimidazole ≥98.0% (by titrimetric analysis)
5-Chloro-2-mercaptobenzimidazole

CAS Number: 25369-78-2
MDL Number: MFCD01658762
Molecular Formula: C7H5ClN2S
Molecular Weight: 184.64
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 294


Other Product Names / Synonyms: 5-Chloro-2-benzimidazolethiol

Formula: C₇H₅ClN₂S
MW: 184.64 g/mol
Melting Pt: 294 °C
MDL Number: MFCD01658762
CAS Number: 25369-78-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff2e55c213ee3c740e22b411814dc17711b8fd4b&vid=06feeb222956d1b6018358d79f1e1577920a0eb2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR