O-(4-Chlorophenyl) chlorothioformate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Chlorothioformic acid O-(4-Chlorophenyl) ester, 4-Chlorophenyl chlorothioformate, 4-Chlorophenyl chlorothionoformate

C1588-25G C1588-5G
TCC1588-25GEA 514.7 USD
TCC1588-25G TCC1588-5G
O-(4-Chlorophenyl) chlorothioformate ≥98.0% (by titrimetric analysis)
O-(4-Chlorophenyl) chlorothioformate

CAS Number: 937-64-4
MDL Number: MFCD00014465
Molecular Formula: C7H4Cl2OS
Molecular Weight: 207.07
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 131
Flash Point (°C): 110
Specific Gravity (20/20): 1.40


Other Product Names / Synonyms: 4-Chlorophenyl Chlorothioformate, Chlorothioformic Acid O-(4-Chlorophenyl) Ester

Formula: C₇H₄Cl₂OS
Boiling Pt: 131 °C
Density: 1.40 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00014465
CAS Number: 937-64-4
UN: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=079b57bded29cce84d0ae8a95aeb6c39c87b39d6&vid=079b57bded29cce84d0ae8a95aeb6c39c87b39d6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR