Methyl chlorodifluoroacetate 97,0
Supplier: TCI America
C1586-25G
C1586-5G
TCC1586-25GEA
59
USD
TCC1586-25G
TCC1586-5G
Methyl chlorodifluoroacetate 97,0
Methyl chlorodifluoroacetate
CAS Number: 1514-87-0
MDL Number: MFCD00000775
Molecular Formula: C3H3ClF2O2
Molecular Weight: 144.50
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 82
Flash Point (°C): 19
Specific Gravity (20/20): 1.35
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks, Difluoromethylating Agents
Other Product Names / Synonyms: Chlorodifluoroacetic Acid Methyl Ester
Formula:
C₃H₃ClF₂O₂ MW: 144.51 Boiling Pt: 82 °C Density: 1.35 (20 °C) Flash Pt: 19 °C |
MDL Number:
MFCD00000775 CAS Number: 1514-87-0 UN: 2924 ADR: 3 II |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...