3-Chlorothiophenol ≥97.0% (by GC)
Supplier: TCI America
CAS Number: 2037-31-2
MDL Number: MFCD00004839
Molecular Formula: C6H5ClS
Molecular Weight: 144.62
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 90
Specific Gravity (20/20): 1.26
Other Product Names / Synonyms: 3-Chloromercaptobenzene, 3-Chlorothiophenol
Formula:
C₆H₅ClS Boiling Pt: 88 °C Density: 1.26 (20 °C) Flash Pt: 90 °C |
MDL Number:
MFCD00004839 CAS Number: 2037-31-2 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...