Methyl oxalyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Methyl 2-chloro-2-oxoacetate, Methyl chloroglyoxylate

C1561-250G C1561-25G
TCC1561-250GEA 443.57 USD
TCC1561-250G TCC1561-25G
Methyl oxalyl chloride ≥98.0% (by GC, titration analysis)
Methyl oxalyl chloride

CAS Number: 5781-53-3
MDL Number: MFCD00000705
Molecular Formula: C3H3ClO3
Molecular Weight: 122.50
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 125
Flash Point (°C): 46
Specific Gravity (20/20): 1.33


Other Product Names / Synonyms: Chloroglyoxylic Acid Methyl Ester, Methyl Oxalyl Chloride

Formula: C₃H₃ClO₃
MW: 122.51 g/mol
Boiling Pt: 125 °C
Density: 1.33 (20 °C)
Flash Pt: 46 °C
MDL Number: MFCD00000705
CAS Number: 5781-53-3
UN: 2920
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a4ad9f806f8ea16030c093adc3b8ae2bf9d06fa&vid=6a4ad9f806f8ea16030c093adc3b8ae2bf9d06fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR