α-Bromo-o-toluonitrile ≥98.0% (by GC)

Supplier: TCI America

C1556-10G C1556-25G
TCC1556-10GEA 63.8 USD
TCC1556-10G TCC1556-25G
α-Bromo-o-toluonitrile ≥98.0% (by GC)
α-Bromo-o-toluonitrile

CAS Number: 22115-41-9
MDL Number: MFCD00001794
Molecular Formula: C8H6BrN
Molecular Weight: 196.05
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 111
Melting point (°C): 72
Flash Point (°C): 110


Other Product Names / Synonyms: 2-(Bromomethyl)benzonitrile, alpha-Bromo-o-tolunitrile

Formula: C₈H₆BrN
Boiling Pt: 111 °C
Melting Pt: 72 °C
Flash Pt: 110 °C
MDL Number: MFCD00001794
CAS Number: 22115-41-9
UN: 2923
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0e87c9cd8bb953159b061b115204431da0df0c1&vid=a0e87c9cd8bb953159b061b115204431da0df0c1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR