(Chloromethylene)dimethylammonium chloride ≥95.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
N-Chloromethylen-N,N-Dimethyl Ammonium Chloride
C1545-250G
C1545-25G
TCC1545-250GEA
793
USD
TCC1545-250G
TCC1545-25G
(Chloromethylene)dimethylammonium chloride ≥95.0% (by titrimetric analysis)
(Chloromethylene)dimethylammonium chloride
CAS Number: 3724-43-4
MDL Number: MFCD00011868
Molecular Formula: C3H7Cl2N
Molecular Weight: 128.00
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Formylation
Other Product Names / Synonyms: Vilsmeier Reagent
Formula:
C₃H₇Cl₂N MW: 128.00 g/mol |
MDL Number:
MFCD00011868 CAS Number: 3724-43-4 |
Specification Test Results
Purity/Analysis Method | >95.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...