(R)-(-)-α-Cyclohexanemethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: (R)-(-)-1-Cyclohexylethylamine, (R)-1-Cyclohexylethylamine

C1541-25G C1541-5G
TCC1541-25GEA 432 USD
TCC1541-25G TCC1541-5G
(R)-(-)-α-Cyclohexanemethylamine ≥98.0% (by GC, titration analysis)
(R)-(-)-α-Cyclohexanemethylamine

CAS Number: 5913-13-3
MDL Number: MFCD00043338
Molecular Formula: C8H17N
Molecular Weight: 127.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 60
Flash Point (°C): 51
Specific Gravity (20/20): 0.87
Specific rotation [a]20/D: -3.3 deg (neat)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-(-)-(1-Aminoethyl)cyclohexane

Formula: C₈H₁₇N
MW: 127.23 g/mol
Boiling Pt: 60 °C
Density: 0.87 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00043338
CAS Number: 5913-13-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e98b3773f87a7a66a56a1c6823f8948e5b832467&vid=e98b3773f87a7a66a56a1c6823f8948e5b832467&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR