5-Chloro-1-pentyne ≥96.0% (by GC)
Supplier: TCI America
C1522-25ML
C1522-5ML
TCC1522-25MLEA
121
USD
TCC1522-25ML
TCC1522-5ML
5-Chloro-1-pentyne ≥96.0% (by GC)
5-Chloro-1-pentyne
CAS Number: 14267-92-6
MDL Number: MFCD00001014
Molecular Formula: C5H7Cl
Molecular Weight: 102.56
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): 15
Specific Gravity (20/20): 0.98
Product Categories: Synthetic Organic Chemistry, Terminal Alkynes (Click Chemistry)
Other Product Names / Synonyms: 4-Pentynyl Chloride
Formula:
C₅H₇Cl MW: 102.56 g/mol Boiling Pt: 113 °C Density: 0.98 (20 °C) Flash Pt: 15 °C |
MDL Number:
MFCD00001014 CAS Number: 14267-92-6 UN: 1993 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >96.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...