4-Chloro-2-fluorotoluene ≥98.0% (by GC)
Supplier: TCI America
C1513-250G
C1513-25G
TCC1513-250GEA
158
USD
TCC1513-250G
TCC1513-25G
4-Chloro-2-fluorotoluene ≥98.0% (by GC)
4-Chloro-2-fluorotoluene
CAS Number: 452-75-5
MDL Number: MFCD00000571
Molecular Formula: C7H6ClF
Molecular Weight: 144.57
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Flash Point (°C): 51
Specific Gravity (20/20): 1.19
Formula:
C₇H₆ClF Density: 1.19 (20 °C) Flash Pt: 51 °C |
MDL Number:
MFCD00000571 CAS Number: 452-75-5 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...