Chlorodifluoroacetic anhydride 98,0

Supplier: TCI America

C1503-10G C1503-25G
TCC1503-10GEA 111 USD
TCC1503-10G TCC1503-25G
Chlorodifluoroacetic anhydride 98,0
Chlorodifluoroacetic anhydride

CAS Number: 2834-23-3
MDL Number: MFCD00000774
Molecular Formula: C4Cl2F4O3
Molecular Weight: 242.93
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid

Formula: C₄Cl₂F₄O₃
MW: 242.94
Density: 1.3953
MDL Number: MFCD00000774
CAS Number: 2834-23-3
UN: 3265
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7766ae3446c0599780b6ffd4e5f2969e281a2a93&vid=7766ae3446c0599780b6ffd4e5f2969e281a2a93&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR