Ethyl chlorodifluoroacetate ≥97.0% (by GC)
Supplier: TCI America
C1458-25G
C1458-500G
TCC1458-25GEA
80
USD
TCC1458-25G
TCC1458-500G
Ethyl chlorodifluoroacetate ≥97.0% (by GC)
Ethyl chlorodifluoroacetate
CAS Number: 383-62-0
MDL Number: MFCD00013662
Molecular Formula: C4H5ClF2O2
Molecular Weight: 158.53
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 96
Flash Point (°C): 20
Specific Gravity (20/20): 1.27
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: Chlorodifluoroacetic Acid Ethyl Ester
Formula:
C₄H₅ClF₂O₂ MW: 158.53 g/mol Boiling Pt: 96 °C Density: 1.27 (20 °C) Flash Pt: 20 °C |
MDL Number:
MFCD00013662 CAS Number: 383-62-0 UN: 2924 ADR: 3,II |
Specification Test Results
Purity/Analysis Method | >97.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...