(1R,2R)-(-)-1,2-Diaminocyclohexane ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(R,R)-DACH, (1R,2R)-Cyclohexane-1,2-diamine
C1447-25G
C1447-5G
TCC1447-25GEA
442.91
USD
TCC1447-25G
TCC1447-5G
(1R,2R)-(-)-1,2-Diaminocyclohexane ≥98.0% (by GC, titration analysis)
(1R,2R)-(-)-1,2-Diaminocyclohexane
CAS Number: 20439-47-8
MDL Number: MFCD00062985
Molecular Formula: C6H14N2
Molecular Weight: 114.19
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Boiling point (°C): 78
Melting point (°C): 43
Flash Point (°C): 70
Specific rotation [a]20/D: -25.5 deg (C=5, 1mol/L HCl)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (1R)-(-)-trans-1,2-Diaminocyclohexane
Formula:
C₆H₁₄N₂ MW: 114.19 g/mol Boiling Pt: 78 °C Melting Pt: 43 °C Flash Pt: 70 °C |
MDL Number:
MFCD00062985 CAS Number: 20439-47-8 UN: 3259 ADR: 8,II |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...